Product Details
- SNP ID
-
rs150441632
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:42142196 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGCCTTCTCCTCAGCTGTTTGTC[A/G]CTCCACACCTGTGGGTGGGGGAAGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLA2G4F
PubMed Links
Gene Details
- Gene
- PLA2G4F
- Gene Name
- phospholipase A2 group IVF
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_213600.3 |
2430 |
Nonsense Mutation |
CGA,TGA |
R780* |
NP_998765.3 |
View Full Product Details