Product Details

SNP ID
rs150510618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90395697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTAACATTTAAATATGTGAGGT[A/C]TGTTCAGTGCTTAGGTCTTGGACAG
Phenotype
MIM: 603379
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IQGAP1 PubMed Links

Gene Details

Gene
IQGAP1
Gene Name
IQ motif containing GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003870.3 Intron NP_003861.1

View Full Product Details