Product Details
- SNP ID
-
rs150851735
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:56921042 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTTGCAGATGTTTTCCCCACCTG[C/T]TAATAGTGGGAAAACTAGACCAACT
- Phenotype
-
MIM: 600480
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC145783
PubMed Links
Gene Details
- Gene
- LOC145783
- Gene Name
- uncharacterized LOC145783
There are no transcripts associated with this gene.
- Gene
- TCF12
- Gene Name
- transcription factor 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001306219.2 |
233 |
Intron |
|
|
NP_001293148.1 |
NM_001306220.2 |
233 |
Intron |
|
|
NP_001293149.1 |
NM_001322151.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_001309080.1 |
NM_001322152.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_001309081.1 |
NM_001322154.1 |
233 |
UTR 5 |
|
|
NP_001309083.1 |
NM_001322156.1 |
233 |
UTR 5 |
|
|
NP_001309085.1 |
NM_001322157.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_001309086.1 |
NM_001322158.1 |
233 |
UTR 5 |
|
|
NP_001309087.1 |
NM_001322159.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_001309088.1 |
NM_001322161.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_001309090.1 |
NM_001322162.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_001309091.1 |
NM_001322164.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_001309093.1 |
NM_001322165.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_001309094.1 |
NM_003205.3 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_003196.1 |
NM_207036.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_996919.1 |
NM_207037.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_996920.1 |
NM_207038.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
NP_996921.1 |
NM_207040.1 |
233 |
Intron |
|
|
NP_996923.1 |
XM_011521959.2 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011520261.1 |
XM_011521960.2 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011520262.1 |
XM_011521961.2 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011520263.1 |
XM_011521962.2 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011520264.1 |
XM_011521963.2 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011520265.1 |
XM_011521965.1 |
233 |
UTR 5 |
|
|
XP_011520267.1 |
XM_011521966.2 |
233 |
Intron |
|
|
XP_011520268.1 |
XM_011521967.1 |
233 |
UTR 5 |
|
|
XP_011520269.1 |
XM_011521969.1 |
233 |
Intron |
|
|
XP_011520271.1 |
XM_017022520.1 |
233 |
Missense Mutation |
GCT,GTT |
A31V |
XP_016878009.1 |
View Full Product Details