Product Details

SNP ID
rs137868909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75477583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTCCATGAGCTCTTCTTTTAAA[C/T]ATGTACTGGGCACTTATAGGCTCTC
Phenotype
MIM: 605294
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST6 PubMed Links

Gene Details

Gene
CHST6
Gene Name
carbohydrate sulfotransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021615.4 3041 UTR 3 NP_067628.1
XM_005255955.4 3041 UTR 3 XP_005256012.1
XM_011523085.2 3041 UTR 3 XP_011521387.1

View Full Product Details