Product Details

SNP ID
rs137903168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84294984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGGGAGGAAATGCCTTTAACC[A/G]GCGTGGGGCCGGGCAGCTGCAGGAG
Phenotype
MIM: 605322
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105376777 PubMed Links

Gene Details

Gene
LOC105376777
Gene Name
uncharacterized LOC105376777
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011523512.2 279 Intron XP_011521814.1
Gene
WFDC1
Gene Name
WAP four-disulfide core domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282466.1 279 Missense Mutation AGC,GGC S5G NP_001269395.1
NM_001282467.1 279 Missense Mutation AGC,GGC S5G NP_001269396.1
NM_021197.3 279 Missense Mutation AGC,GGC S5G NP_067020.2
XM_005256079.3 279 Missense Mutation AGC,GGC S5G XP_005256136.1

View Full Product Details