Product Details

SNP ID
rs138324633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74301140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGTATGGAATGTTTAAGAAAGT[C/G]AATGGTATGTCTTGATGTTCTAAGG
Phenotype
MIM: 157970
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PSMD7 PubMed Links

Gene Details

Gene
PSMD7
Gene Name
proteasome 26S subunit, non-ATPase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002811.4 395 Silent Mutation GTC,GTG V85V NP_002802.2

View Full Product Details