Product Details

SNP ID
rs138372665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89561640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTCTGTGGATCCGAGGAGGCG[A/G]AACAAGTCCACGGAGTCCCTGCAGG
Phenotype
MIM: 113703 MIM: 602783
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL13 PubMed Links

Gene Details

Gene
RPL13
Gene Name
ribosomal protein L13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000977.3 385 Silent Mutation CGA,CGG R103R NP_000968.2
NM_001243130.1 385 Silent Mutation CGA,CGG R84R NP_001230059.1
NM_001243131.1 385 Intron NP_001230060.1
NM_033251.2 385 Silent Mutation CGA,CGG R103R NP_150254.1
Gene
SNORD68
Gene Name
small nucleolar RNA, C/D box 68
There are no transcripts associated with this gene.

Gene
SPG7
Gene Name
SPG7, paraplegin matrix AAA peptidase subunit
There are no transcripts associated with this gene.

View Full Product Details