Product Details

SNP ID
rs139088208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3283982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGCTTCCAAGAGGCAGCTGGTC[C/T]CCGGGAAGCCCTCAGCCGGCTCCAA
Phenotype
MIM: 604191
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF263 PubMed Links

Gene Details

Gene
ZNF263
Gene Name
zinc finger protein 263
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005741.4 223 Missense Mutation CCC,CTC P55L NP_005732.2
XM_006720831.1 223 Missense Mutation CCC,CTC P55L XP_006720894.1
XM_006720832.1 223 Missense Mutation CCC,CTC P55L XP_006720895.1
XM_006720833.1 223 Missense Mutation CCC,CTC P55L XP_006720896.1
XM_006720834.3 223 UTR 5 XP_006720897.1
XM_011522344.1 223 Missense Mutation CCC,CTC P55L XP_011520646.1
XM_017022850.1 223 UTR 5 XP_016878339.1

View Full Product Details