Product Details

SNP ID
rs139411224
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69109615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCACCGGCGCATGCGACGTGCC[A/G]GCCAGGCCCTGAAGCTGCCCTCCCC
Phenotype
MIM: 613202 MIM: 602428
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CHTF8 PubMed Links
Additional Information
For this assay, SNP(s) [rs2232228] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CHTF8
Gene Name
chromosome transmission fidelity factor 8
There are no transcripts associated with this gene.

Gene
HAS3
Gene Name
hyaluronan synthase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199280.1 311 Missense Mutation AGC,GGC S74G NP_001186209.1
NM_005329.2 311 Missense Mutation AGC,GGC S74G NP_005320.2
NM_138612.2 311 Missense Mutation AGC,GGC S74G NP_619515.1
XM_005255921.2 311 Missense Mutation AGC,GGC S74G XP_005255978.1
XM_011523061.2 311 Missense Mutation AGC,GGC S74G XP_011521363.1

View Full Product Details