Product Details

SNP ID
rs139452648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84478537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAGGTCAAAGCTTACAGCAAAA[A/G]ACAAAAGGAAGAGGTATATATCTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLDC1 PubMed Links

Gene Details

Gene
TLDC1
Gene Name
TBC/LysM-associated domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020947.3 3151 UTR 3 NP_065998.3
XM_005256074.3 3151 UTR 3 XP_005256131.1
XM_005256075.2 3151 UTR 3 XP_005256132.1
XM_006721240.2 3151 UTR 3 XP_006721303.1
XM_011523248.2 3151 UTR 3 XP_011521550.1
XM_011523249.2 3151 Intron XP_011521551.1
XM_017023510.1 3151 UTR 3 XP_016878999.1
XM_017023511.1 3151 UTR 3 XP_016879000.1
XM_017023512.1 3151 UTR 3 XP_016879001.1

View Full Product Details