Product Details

SNP ID
rs139579879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84222453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGGAAGGCCATGATGAGGATCC[C/T]GCTCAGGATGCTGCTGAGGGCCACC
Phenotype
MIM: 607603
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNG4 PubMed Links

Gene Details

Gene
KCNG4
Gene Name
potassium voltage-gated channel modifier subfamily G member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172347.2 1445 Missense Mutation AGG,GGG R442G NP_758857.1

View Full Product Details