Product Details

SNP ID
rs139782780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57996748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGGGCTTCTCGCCTGTGTGTAC[C/T]CGCCGGTGCCGCTGCAGGTCTGACG
Phenotype
MIM: 610264 MIM: 613276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEPP PubMed Links

Gene Details

Gene
TEPP
Gene Name
testis, prostate and placenta expressed
There are no transcripts associated with this gene.

Gene
USB1
Gene Name
U6 snRNA biogenesis phosphodiesterase 1
There are no transcripts associated with this gene.

Gene
ZNF319
Gene Name
zinc finger protein 319
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020807.2 2141 Silent Mutation NP_065858.1
XM_005256069.3 2141 Silent Mutation XP_005256126.1

View Full Product Details