Product Details

SNP ID
rs139806180
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2971165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTCTCCCTCCCTAGGGCTGGCC[C/T]TGCTGGGCTTCCTGGTGCTGGTGCC
Phenotype
MIM: 609899 MIM: 614578 MIM: 602474
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KREMEN2 PubMed Links
Additional Information
For this assay, SNP(s) [rs4786365] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KREMEN2
Gene Name
kringle containing transmembrane protein 2
There are no transcripts associated with this gene.

Gene
PAQR4
Gene Name
progestin and adipoQ receptor family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284511.1 605 Intron NP_001271440.1
NM_001284512.1 605 Intron NP_001271441.1
NM_001284513.1 605 UTR 5 NP_001271442.1
NM_001324118.1 605 Silent Mutation CTG,TTG L59L NP_001311047.1
NM_152341.4 605 Silent Mutation CTG,TTG L59L NP_689554.2
Gene
PKMYT1
Gene Name
protein kinase, membrane associated tyrosine/threonine 1
There are no transcripts associated with this gene.

View Full Product Details