Product Details

SNP ID
rs140902087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3090486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAAGGGACCCATCTTCACCAGG[A/G]CCGCTAGCCGCAGCGCCCCGTCCGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN10 PubMed Links

Gene Details

Gene
ZSCAN10
Gene Name
zinc finger and SCAN domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282415.1 875 UTR 5 NP_001269344.1
NM_001282416.1 875 Silent Mutation GGC,GGT G179G NP_001269345.1
NM_032805.2 875 Silent Mutation GGC,GGT G316G NP_116194.2
XM_017023791.1 875 Silent Mutation GGC,GGT G316G XP_016879280.1
XM_017023792.1 875 Silent Mutation GGC,GGT G316G XP_016879281.1

View Full Product Details