Product Details

SNP ID
rs141008024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1494475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCACTTCTCGCCTGTCCTCAGAT[C/G]TCTTGCCAGCAGGCTGAGCCCAGCC
Phenotype
MIM: 613442 MIM: 611140
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTX4 PubMed Links

Gene Details

Gene
PTX4
Gene Name
pentraxin 4
There are no transcripts associated with this gene.

Gene
TELO2
Gene Name
telomere maintenance 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016111.3 544 Missense Mutation TCT,TGT S65C NP_057195.2
XM_011522773.2 544 Missense Mutation TCT,TGT S65C XP_011521075.1
XM_011522774.2 544 Missense Mutation TCT,TGT S65C XP_011521076.1
XM_011522775.2 544 Missense Mutation TCT,TGT S65C XP_011521077.1
XM_011522776.2 544 Missense Mutation TCT,TGT S65C XP_011521078.1
XM_011522777.2 544 Missense Mutation TCT,TGT S65C XP_011521079.1
XM_011522778.2 544 Missense Mutation TCT,TGT S65C XP_011521080.1
XM_017023914.1 544 Missense Mutation TCT,TGT S65C XP_016879403.1

View Full Product Details