Product Details
- SNP ID
-
rs141300146
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89738217 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCGAGCCCCTCTGTGACCACAGAG[G/T]GCCAGGCGGTGAAGCCCGAACCCAC
- Phenotype
-
MIM: 607139
MIM: 608460
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FANCA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17227424] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FANCA
- Gene Name
- Fanconi anemia complementation group A
- Gene
- ZNF276
- Gene Name
- zinc finger protein 276
View Full Product Details