Product Details

SNP ID
rs141625180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70481396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGATGGTGGCCATCTGGGAGAGC[C/T]GGGCAAACTTGTCTCGGATGGTCCA
Phenotype
MIM: 606976 MIM: 608675
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COG4 PubMed Links

Gene Details

Gene
COG4
Gene Name
component of oligomeric golgi complex 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195139.1 2369 Missense Mutation CAG,CGG Q712R NP_001182068.1
NM_015386.2 2369 Missense Mutation CAG,CGG Q733R NP_056201.2
XM_011522981.2 2369 Missense Mutation CAG,CGG Q591R XP_011521283.1
Gene
FUK
Gene Name
fucokinase
There are no transcripts associated with this gene.

View Full Product Details