Product Details

SNP ID
rs142008098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84194090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGGAAGTGCTCAGAGCCTTCC[G/T]CCTGAGAACAGGGTGGCGTGGGCTC
Phenotype
MIM: 604905
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAD2 PubMed Links

Gene Details

Gene
ADAD2
Gene Name
adenosine deaminase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145400.1 312 Intron NP_001138872.1
NM_139174.3 312 Missense Mutation CGC,CTC R168L NP_631913.3
XM_005255814.1 312 Intron XP_005255871.1
XM_011522919.1 312 Missense Mutation CGC,CTC R168L XP_011521221.1
XM_011522920.1 312 Missense Mutation CGC,CTC R38L XP_011521222.1
Gene
TAF1C
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit C
There are no transcripts associated with this gene.

View Full Product Details