Product Details

SNP ID
rs142521005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29873379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCTGGTAGTGGTGCTTGTACAG[C/T]GTCTGGTAGCCATTCTCGGGAACCC
Phenotype
MIM: 616667
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDIPT-AS1 PubMed Links

Gene Details

Gene
CDIPT-AS1
Gene Name
CDIPT antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
SEZ6L2
Gene Name
seizure related 6 homolog like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114099.2 2351 Silent Mutation ACA,ACG T713T NP_001107571.1
NM_001114100.2 2351 Silent Mutation ACA,ACG T669T NP_001107572.1
NM_001243332.1 2351 Silent Mutation ACA,ACG T783T NP_001230261.1
NM_001243333.1 2351 Silent Mutation ACA,ACG T739T NP_001230262.1
NM_012410.3 2351 Silent Mutation ACA,ACG T713T NP_036542.1
NM_201575.3 2351 Silent Mutation ACA,ACG T783T NP_963869.2
XM_005255252.2 2351 Intron XP_005255309.1
XM_017023135.1 2351 Silent Mutation ACA,ACG T599T XP_016878624.1

View Full Product Details