Product Details

SNP ID
rs143001723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:399013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGGCCCTCCTGGACCCGGGAG[A/C]GGACCCTGGTGGCGGTGAAGCCCGA
Phenotype
MIM: 615839 MIM: 601818
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DECR2 PubMed Links

Gene Details

Gene
DECR2
Gene Name
2,4-dienoyl-CoA reductase 2, peroxisomal
There are no transcripts associated with this gene.

Gene
LOC100134368
Gene Name
uncharacterized LOC100134368
There are no transcripts associated with this gene.

Gene
NME4
Gene Name
NME/NM23 nucleoside diphosphate kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286433.1 146 Silent Mutation AGG,CGG R39R NP_001273362.1
NM_001286435.1 146 Silent Mutation AGG,CGG R39R NP_001273364.1
NM_001286436.1 146 UTR 5 NP_001273365.1
NM_001286438.1 146 UTR 5 NP_001273367.1
NM_001286439.1 146 UTR 5 NP_001273368.1
NM_001286440.1 146 UTR 5 NP_001273369.1
NM_005009.2 146 Silent Mutation AGG,CGG R39R NP_005000.1

View Full Product Details