Product Details

SNP ID
rs143347015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4851975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATTTATGTATAATATCAAGAG[C/T]GCTTACAAGTCAATAAGAAAGTCAC
Phenotype
MIM: 610660 MIM: 609771
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLYR1 PubMed Links

Gene Details

Gene
GLYR1
Gene Name
glyoxylate reductase 1 homolog
There are no transcripts associated with this gene.

Gene
UBN1
Gene Name
ubinuclein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079514.2 Intron NP_001072982.1
NM_001288656.1 Intron NP_001275585.1
XM_005255277.3 Intron XP_005255334.1
XM_005255278.3 Intron XP_005255335.1
XM_005255279.3 Intron XP_005255336.1
XM_011522465.1 Intron XP_011520767.1
XM_011522466.2 Intron XP_011520768.1
XM_011522467.1 Intron XP_011520769.1
XM_017023189.1 Intron XP_016878678.1
XM_017023190.1 Intron XP_016878679.1
XM_017023191.1 Intron XP_016878680.1
XM_017023192.1 Intron XP_016878681.1

View Full Product Details