Product Details

SNP ID
rs143591002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84567481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTATCAAAGTGATGTCACCATAC[A/G]TGGAGGTGGAAGGAGACAGGCGTAA
Phenotype
MIM: 606748
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COTL1 PubMed Links

Gene Details

Gene
COTL1
Gene Name
coactosin like F-actin binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021149.3 Intron NP_066972.1

View Full Product Details