Product Details

SNP ID
rs143840454
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29780254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAAGCTGGTATTTGTGACAGA[C/G]AAGGGCCGCTATCTGTCTTTTGGGA
Phenotype
MIM: 603213
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
KIF22 PubMed Links

Gene Details

Gene
KIF22
Gene Name
kinesin family member 22
There are no transcripts associated with this gene.

Gene
ZG16
Gene Name
zymogen granule protein 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152338.3 492 Missense Mutation GAC,GAG D113E NP_689551.2
XM_011545921.2 492 Missense Mutation GAC,GAG D113E XP_011544223.1

View Full Product Details