Product Details

SNP ID
rs143894366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29911050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGGCGATTTTGCGGCCCTGCCC[A/G]TAGAAAGACCAGCAGCAGATCTCGT
Phenotype
MIM: 608947
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASPHD1 PubMed Links

Gene Details

Gene
ASPHD1
Gene Name
aspartate beta-hydroxylase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181718.3 968 Intron NP_859069.2
XM_017023107.1 968 Intron XP_016878596.1
Gene
KCTD13
Gene Name
potassium channel tetramerization domain containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178863.4 968 Silent Mutation TAC,TAT Y227Y NP_849194.1
XM_011545783.2 968 Silent Mutation TAC,TAT Y245Y XP_011544085.1
XM_011545784.2 968 Silent Mutation TAC,TAT Y100Y XP_011544086.1
XM_017023104.1 968 Silent Mutation TAC,TAT Y76Y XP_016878593.1
XM_017023105.1 968 Intron XP_016878594.1
XM_017023106.1 968 Intron XP_016878595.1

View Full Product Details