Product Details

SNP ID
rs143936171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4746470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAGTCCCCCACCATCTTTATTG[A/G]CCTGCGGCAGATGGAGCTACCAGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf71 PubMed Links

Gene Details

Gene
C16orf71
Gene Name
chromosome 16 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139170.2 1330 Missense Mutation GAC,GGC D380G NP_631909.2
XM_005255144.3 1330 Missense Mutation GAC,GGC D242G XP_005255201.1
XM_017022975.1 1330 Missense Mutation GAC,GGC D403G XP_016878464.1
XM_017022976.1 1330 Missense Mutation GAC,GGC D380G XP_016878465.1
Gene
ZNF500
Gene Name
zinc finger protein 500
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303450.1 1330 Intron NP_001290379.1
NM_021646.3 1330 Intron NP_067678.1
XM_005255243.3 1330 Intron XP_005255300.1
XM_011522453.2 1330 Intron XP_011520755.1
XM_011522454.2 1330 Intron XP_011520756.1
XM_017023121.1 1330 Intron XP_016878610.1

View Full Product Details