Product Details

SNP ID
rs144013707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:372248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGCCCAGCGCTGCCACGAGGT[A/G]GGGTAGCACTGGCGCCGGTGCCCGC
Phenotype
MIM: 604853
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100134368 PubMed Links

Gene Details

Gene
LOC100134368
Gene Name
uncharacterized LOC100134368
There are no transcripts associated with this gene.

Gene
MRPL28
Gene Name
mitochondrial ribosomal protein L28
There are no transcripts associated with this gene.

Gene
TMEM8A
Gene Name
transmembrane protein 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021259.2 2184 Silent Mutation CCC,CCT P685P NP_067082.2

View Full Product Details