Product Details

SNP ID
rs144183945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31037977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTCATCAACAAGTGCAATTCA[A/G]TGCAGTCCGAATACCGGGAGAAGAA
Phenotype
MIM: 186591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928762 PubMed Links

Gene Details

Gene
LOC101928762
Gene Name
uncharacterized LOC101928762
There are no transcripts associated with this gene.

Gene
STX4
Gene Name
syntaxin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272095.1 881 Missense Mutation ATG,GTG M66V NP_001259024.1
NM_001272096.1 881 Missense Mutation ATG,GTG M142V NP_001259025.1
NM_004604.4 881 Missense Mutation ATG,GTG M144V NP_004595.2
XM_005255521.1 881 Missense Mutation ATG,GTG M140V XP_005255578.1
XM_011545925.2 881 Missense Mutation ATG,GTG M1V XP_011544227.1

View Full Product Details