Product Details
- SNP ID
-
rs144188544
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:28608326 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATCCAACAGAGTCTGGGGGAGCA[C/G]AGCCAGGGGCAGGTGTGTCTTCAGG
- Phenotype
-
MIM: 171150
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SULT1A1
PubMed Links
- Additional Information
-
The SULT1A1 gene exhibits copy number variation. Individuals may carry deletion alleles or extra copies of SULT1A1. SULT1A1 SNP genotyping assays run on samples having 2 or more gene copies that are homozygous for the SNP allele will cluster together, and samples having more than 2 gene copies that are heterozygous may run between the 2 copy heterozygous and homozygous clusters. For accurate SULT1A1 genotype analysis, copy number analysis must be done. For more information, refer to the PGx Experiments User Guide (Pub. # MAN0009612) Chapter 2 Copy Number Variation section.
Gene Details
- Gene
- SULT1A1
- Gene Name
- sulfotransferase family 1A member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001055.3 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
NP_001046.2 |
NM_177529.2 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
NP_803565.1 |
NM_177530.2 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
NP_803566.1 |
NM_177534.2 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
NP_803878.1 |
NM_177536.3 |
767 |
Intron |
|
|
NP_803880.1 |
XM_017023604.1 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
XP_016879093.1 |
XM_017023605.1 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
XP_016879094.1 |
XM_017023606.1 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
XP_016879095.1 |
XM_017023607.1 |
767 |
Silent Mutation |
CTG,GTG |
L204V |
XP_016879096.1 |
XM_017023608.1 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
XP_016879097.1 |
XM_017023609.1 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
XP_016879098.1 |
XM_017023610.1 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
XP_016879099.1 |
XM_017023611.1 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
XP_016879100.1 |
XM_017023612.1 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
XP_016879101.1 |
XM_017023613.1 |
767 |
Silent Mutation |
CTG,GTG |
L113V |
XP_016879102.1 |
View Full Product Details