Product Details

SNP ID
rs144794688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67930002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACTCGAGTATACACAGCAGGTG[C/T]GCGCACATTGCAGTTTTTGGTGCCC
Phenotype
MIM: 118888 MIM: 606967 MIM: 177015 MIM: 176847
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTRL PubMed Links

Gene Details

Gene
CTRL
Gene Name
chymotrypsin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001907.2 750 Missense Mutation ACA,GCA T243A NP_001898.1
Gene
LCAT
Gene Name
lecithin-cholesterol acyltransferase
There are no transcripts associated with this gene.

Gene
PSKH1
Gene Name
protein serine kinase H1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006742.2 750 Intron NP_006733.1
Gene
PSMB10
Gene Name
proteasome subunit beta 10
There are no transcripts associated with this gene.

View Full Product Details