Product Details

SNP ID
rs144882405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30086820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGGCTGCTGGCTCTGGGCCCC[A/G]CAGTTTCCTCTTCACACGGGCGTCT
Phenotype
MIM: 602035 MIM: 602427 MIM: 609724
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928595 PubMed Links

Gene Details

Gene
LOC101928595
Gene Name
uncharacterized LOC101928595
There are no transcripts associated with this gene.

Gene
PPP4C
Gene Name
protein phosphatase 4 catalytic subunit
There are no transcripts associated with this gene.

Gene
TBX6
Gene Name
T-box 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004608.3 956 Missense Mutation CGG,TGG R291W NP_004599.2
XM_005255523.2 956 Missense Mutation CGG,TGG R291W XP_005255580.1
XM_011545926.2 956 Missense Mutation CGG,TGG R291W XP_011544228.1
XM_017023614.1 956 Missense Mutation CGG,TGG R291W XP_016879103.1
Gene
YPEL3
Gene Name
yippee like 3
There are no transcripts associated with this gene.

View Full Product Details