Product Details

SNP ID
rs145030670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66579657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAAGCCAGAGCCAGCACCAGCTC[A/C]ACCTCCACCCGGGGCCAAACCCGAG
Phenotype
MIM: 607884 MIM: 607885
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CKLF-CMTM1 PubMed Links

Gene Details

Gene
CKLF-CMTM1
Gene Name
CKLF-CMTM1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202509.1 210 Intron NP_001189438.1
NM_001204098.1 210 Intron NP_001191027.1
NM_001204099.1 210 Intron NP_001191028.1
Gene
CMTM1
Gene Name
CKLF like MARVEL transmembrane domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052999.3 210 Intron NP_443725.3
NM_181268.2 210 Intron NP_851785.2
NM_181269.2 210 Intron NP_851786.1
NM_181270.2 210 Intron NP_851787.1
NM_181271.2 210 Intron NP_851788.1
NM_181272.2 210 Intron NP_851789.1
NM_181283.2 210 Intron NP_851800.1
NM_181296.2 210 Intron NP_851813.1
Gene
CMTM2
Gene Name
CKLF like MARVEL transmembrane domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199317.1 210 Missense Mutation CAA,CCA Q17P NP_001186246.1
NM_144673.2 210 Missense Mutation CAA,CCA Q17P NP_653274.1

View Full Product Details