Product Details
- SNP ID
-
rs145134196
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:67675347 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTGCGGTCCCAGGTGCGGCGGTC[A/G]CCCTGCCCCTGGAAGGACTGGCGCA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C16orf86
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201012397] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C16orf86
- Gene Name
- chromosome 16 open reading frame 86
There are no transcripts associated with this gene.
- Gene
- ENKD1
- Gene Name
- enkurin domain containing 1
There are no transcripts associated with this gene.
- Gene
- GFOD2
- Gene Name
- glucose-fructose oxidoreductase domain containing 2
View Full Product Details