Product Details

SNP ID
rs145827882
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88804600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATCCCAGCCTGCCCTTCTCCGG[A/G]CCAGAAGATAAAGAAATCCACCCCG
Phenotype
MIM: 102600 MIM: 605525 MIM: 612222
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
APRT PubMed Links
Additional Information
For this assay, SNP(s) [rs475667] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
APRT
Gene Name
adenine phosphoribosyltransferase
There are no transcripts associated with this gene.

Gene
CDT1
Gene Name
chromatin licensing and DNA replication factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030928.3 338 Missense Mutation GAC,GGC D95G NP_112190.2
Gene
GALNS
Gene Name
galactosamine (N-acetyl)-6-sulfatase
There are no transcripts associated with this gene.

View Full Product Details