Product Details

SNP ID
rs146254840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87698967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCATGTTTCACCTGTGCACC[C/T]GAGGCCAGGAGGAAGTGGTGGCATC
Phenotype
MIM: 605268
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JPH3 PubMed Links

Gene Details

Gene
JPH3
Gene Name
junctophilin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271604.2 Intron NP_001258533.1
NM_001271605.1 Intron NP_001258534.1
NM_020655.3 Intron NP_065706.2
XM_017023483.1 Intron XP_016878972.1
XM_017023484.1 Intron XP_016878973.1
Gene
KLHDC4
Gene Name
kelch domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184854.1 Intron NP_001171783.1
NM_001184856.1 Intron NP_001171785.1
NM_017566.3 Intron NP_060036.2
XM_005255994.3 Intron XP_005256051.1
XM_005255999.1 Intron XP_005256056.1
XM_006721201.1 Intron XP_006721264.1
XM_006721202.1 Intron XP_006721265.1
XM_006721203.2 Intron XP_006721266.1
XM_006721204.3 Intron XP_006721267.1
XM_017023344.1 Intron XP_016878833.1
XM_017023345.1 Intron XP_016878834.1
Gene
LOC100129215
Gene Name
uncharacterized LOC100129215
There are no transcripts associated with this gene.

Gene
LOC105371397
Gene Name
uncharacterized LOC105371397
There are no transcripts associated with this gene.

View Full Product Details