Product Details

SNP ID
rs146702066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87696591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTCTCTTCCAGGGCTCAGCGCC[C/T]ATCCTGGTGGTCATGGTGATCTTGC
Phenotype
MIM: 605268
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JPH3 PubMed Links

Gene Details

Gene
JPH3
Gene Name
junctophilin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271604.2 2438 Intron NP_001258533.1
NM_001271605.1 2438 Intron NP_001258534.1
NM_020655.3 2438 Silent Mutation CCC,CCT P726P NP_065706.2
XM_017023483.1 2438 Intron XP_016878972.1
XM_017023484.1 2438 Intron XP_016878973.1
Gene
KLHDC4
Gene Name
kelch domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184854.1 2438 Intron NP_001171783.1
NM_001184856.1 2438 Intron NP_001171785.1
NM_017566.3 2438 Intron NP_060036.2
XM_005255994.3 2438 Intron XP_005256051.1
XM_005255999.1 2438 Intron XP_005256056.1
XM_006721201.1 2438 Intron XP_006721264.1
XM_006721202.1 2438 Intron XP_006721265.1
XM_006721203.2 2438 Intron XP_006721266.1
XM_006721204.3 2438 UTR 3 XP_006721267.1
XM_017023344.1 2438 Intron XP_016878833.1
XM_017023345.1 2438 Intron XP_016878834.1
Gene
LOC100129215
Gene Name
uncharacterized LOC100129215
There are no transcripts associated with this gene.

Gene
LOC105371397
Gene Name
uncharacterized LOC105371397
There are no transcripts associated with this gene.

View Full Product Details