Product Details

SNP ID
rs147359915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:634777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCCTCCAGGAGGGCTGTGTCC[C/T]GAAGCCCCCATTCTGGGTTCCTGCA
Phenotype
MIM: 608021
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100287175 PubMed Links

Gene Details

Gene
LOC100287175
Gene Name
uncharacterized LOC100287175
There are no transcripts associated with this gene.

Gene
MCRIP2
Gene Name
MAPK regulated corepressor interacting protein 2
There are no transcripts associated with this gene.

Gene
METTL26
Gene Name
methyltransferase like 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040160.2 784 Silent Mutation TCA,TCG S147S NP_001035250.1
NM_001040161.2 784 Missense Mutation CAG,CGG Q73R NP_001035251.1
NM_001040162.2 784 Missense Mutation CAG,CGG Q93R NP_001035252.1
NM_001040165.2 784 Silent Mutation TCA,TCG S127S NP_001035255.1
NM_001288710.1 784 Missense Mutation CAG,CGG Q150R NP_001275639.1
NM_032366.4 784 Missense Mutation CAG,CGG Q170R NP_115742.3
XM_011522713.2 784 Missense Mutation CAG,CGG Q237R XP_011521015.1
XM_011522714.2 784 Missense Mutation AGG,GGG R179G XP_011521016.1
Gene
RAB40C
Gene Name
RAB40C, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
WFIKKN1
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053284.2 784 Intron NP_444514.1

View Full Product Details