Product Details
- SNP ID
-
rs148293871
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:75207128 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGAGGGCCCAGCAGGAGAGGAGC[A/C]AGAGGAAAGCCATGGTGCCGCTGGC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CTRB2
PubMed Links
Gene Details
- Gene
- CTRB2
- Gene Name
- chymotrypsinogen B2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001025200.3 |
47 |
Missense Mutation |
TGG,TTG |
W5L |
NP_001020371.3 |
View Full Product Details