Product Details
- SNP ID
-
rs148332563
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89268637 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCACGGCGTTCAGGGCCGCGGCCTC[A/G]TGCTGCTGCCGCATGAGGAGGCAAG
- Phenotype
-
MIM: 611192
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD11
PubMed Links
Gene Details
- Gene
- ANKRD11
- Gene Name
- ankyrin repeat domain 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256182.1 |
8071 |
Silent Mutation |
CAC,CAT |
H2611H |
NP_001243111.1 |
NM_001256183.1 |
8071 |
Silent Mutation |
CAC,CAT |
H2611H |
NP_001243112.1 |
NM_013275.5 |
8071 |
Silent Mutation |
CAC,CAT |
H2611H |
NP_037407.4 |
XM_006721181.1 |
8071 |
Silent Mutation |
CAC,CAT |
H2577H |
XP_006721244.1 |
XM_006721184.2 |
8071 |
Silent Mutation |
CAC,CAT |
H2512H |
XP_006721247.1 |
XM_011523051.2 |
8071 |
Silent Mutation |
CAC,CAT |
H2611H |
XP_011521353.1 |
XM_011523053.2 |
8071 |
Silent Mutation |
CAC,CAT |
H2611H |
XP_011521355.1 |
XM_011523054.2 |
8071 |
Silent Mutation |
CAC,CAT |
H2577H |
XP_011521356.1 |
XM_011523055.2 |
8071 |
Silent Mutation |
CAC,CAT |
H2577H |
XP_011521357.1 |
XM_011523056.2 |
8071 |
Silent Mutation |
CAC,CAT |
H2568H |
XP_011521358.1 |
XM_011523057.2 |
8071 |
Intron |
|
|
XP_011521359.1 |
XM_017023182.1 |
8071 |
Silent Mutation |
CAC,CAT |
H2611H |
XP_016878671.1 |
XM_017023183.1 |
8071 |
Silent Mutation |
CAC,CAT |
H2611H |
XP_016878672.1 |
XM_017023184.1 |
8071 |
Silent Mutation |
CAC,CAT |
H2611H |
XP_016878673.1 |
XM_017023185.1 |
8071 |
Silent Mutation |
CAC,CAT |
H2611H |
XP_016878674.1 |
XM_017023186.1 |
8071 |
Silent Mutation |
CAC,CAT |
H2611H |
XP_016878675.1 |
XM_017023187.1 |
8071 |
Silent Mutation |
CAC,CAT |
H2611H |
XP_016878676.1 |
View Full Product Details