Product Details
- SNP ID
-
rs148798647
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89179544 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCCCCGATCAGCGTATCCGAGAA[C/T]CACAAGCGTCTCCCCTACCCCCTGG
- Phenotype
-
MIM: 114019
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CDH15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2287358] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDH15
- Gene Name
- cadherin 15
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004933.2 |
248 |
Silent Mutation |
AAC,AAT |
N57N |
NP_004924.1 |
View Full Product Details