Product Details

SNP ID
rs148802894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84294995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGCCTTTAACCGGCGTGGGGCC[A/G]GGCAGCTGCAGGAGGCAGATCATCC
Phenotype
MIM: 605322
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105376777 PubMed Links

Gene Details

Gene
LOC105376777
Gene Name
uncharacterized LOC105376777
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011523512.2 290 Intron XP_011521814.1
Gene
WFDC1
Gene Name
WAP four-disulfide core domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282466.1 290 Silent Mutation CCA,CCG P8P NP_001269395.1
NM_001282467.1 290 Silent Mutation CCA,CCG P8P NP_001269396.1
NM_021197.3 290 Silent Mutation CCA,CCG P8P NP_067020.2
XM_005256079.3 290 Silent Mutation CCA,CCG P8P XP_005256136.1

View Full Product Details