Product Details

SNP ID
rs149294100
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29780155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCGACTATGTGGGTGGTCGCAA[C/G]GGAGACCTGGAGGAGATCTTTCTGC
Phenotype
MIM: 603213
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
KIF22 PubMed Links

Gene Details

Gene
KIF22
Gene Name
kinesin family member 22
There are no transcripts associated with this gene.

Gene
ZG16
Gene Name
zymogen granule protein 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152338.3 393 Silent Mutation AAC,AAG N80K NP_689551.2
XM_011545921.2 393 Missense Mutation AAC,AAG N80K XP_011544223.1

View Full Product Details