Product Details

SNP ID
rs150537360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71448747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGGAGTTACATCTGAAGGCTTT[C/G]CCGCATTCATTACACTCATAGGGTT
Phenotype
MIM: 194527
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF23 PubMed Links

Gene Details

Gene
ZNF23
Gene Name
zinc finger protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304492.1 2101 Intron NP_001291421.1
NM_001304493.1 2101 Silent Mutation GGC,GGG G368G NP_001291422.1
NM_001304494.1 2101 Intron NP_001291423.1
NM_145911.2 2101 Silent Mutation GGC,GGG G426G NP_666016.1

View Full Product Details