Product Details

SNP ID
rs150725455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27226974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTTTTCACCAACCCCAGCCAA[C/T]GGGCATCAGCCACATCCCCCTGGCA
Phenotype
MIM: 611917
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM8 PubMed Links

Gene Details

Gene
KDM8
Gene Name
lysine demethylase 8
There are no transcripts associated with this gene.

Gene
NSMCE1
Gene Name
NSE1 homolog, SMC5-SMC6 complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145080.3 Intron NP_659547.2
XM_006721023.3 Intron XP_006721086.1
XM_011545761.2 Intron XP_011544063.1
XM_011545762.1 Intron XP_011544064.1
XM_017023040.1 Intron XP_016878529.1
XM_017023041.1 Intron XP_016878530.1
XM_017023042.1 Intron XP_016878531.1

View Full Product Details