Product Details
- SNP ID
-
rs150799758
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:81036146 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTGTGCACCGTGCGCGGCTGCGG[C/G]AAGATCCTGCCCAACAGCCCCGCGC
- Phenotype
-
MIM: 614693
MIM: 611509
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATMIN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142522981] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATMIN
- Gene Name
- ATM interactor
- Gene
- CENPN
- Gene Name
- centromere protein N
There are no transcripts associated with this gene.
View Full Product Details