Product Details

SNP ID
rs150935746
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70355444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCCCACATTTTTGGTGGTTCTC[A/C]TATTTGCCTTTCTAATTATGACAAT
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DDX19A PubMed Links

Gene Details

Gene
DDX19A
Gene Name
DEAD-box helicase 19A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320522.1 Intron NP_001307451.1
NM_001320525.1 Intron NP_001307454.1
NM_001320526.1 Intron NP_001307455.1
NM_001320527.1 Intron NP_001307456.1
NM_018332.4 Intron NP_060802.1
Gene
LOC100506083
Gene Name
uncharacterized LOC100506083
There are no transcripts associated with this gene.

View Full Product Details