Product Details
- SNP ID
-
rs150987871
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89179435 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCAGAGCCTCTGCCTGTCTTTGG[C/G]GGTTCCTGGATGGAGGAGGCCCACC
- Phenotype
-
MIM: 114019
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CDH15
PubMed Links
Gene Details
- Gene
- CDH15
- Gene Name
- cadherin 15
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004933.2 |
139 |
Missense Mutation |
GCG,GGG |
A21G |
NP_004924.1 |
View Full Product Details