Product Details

SNP ID
rs151003754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67675473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCACTGCCAGCGAGTCCCTCAA[A/G]AGCAGCTCCTCTTGCGTGGCAGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf86 PubMed Links

Gene Details

Gene
C16orf86
Gene Name
chromosome 16 open reading frame 86
There are no transcripts associated with this gene.

Gene
ENKD1
Gene Name
enkurin domain containing 1
There are no transcripts associated with this gene.

Gene
GFOD2
Gene Name
glucose-fructose oxidoreductase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243650.1 878 Intron NP_001230579.1
NM_030819.3 878 Silent Mutation CTC,CTT L280L NP_110446.3
XM_006721288.3 878 Silent Mutation CTC,CTT L175L XP_006721351.1
XM_017023738.1 878 Silent Mutation CTC,CTT L280L XP_016879227.1

View Full Product Details