Product Details

SNP ID
rs137870769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:73285736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCAGGGGAGGGGCCGCCACGGC[A/G]TACGGGGGAGCCTGGGTGATGGTGC
Phenotype
MIM: 605468
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC42EP4 PubMed Links

Gene Details

Gene
CDC42EP4
Gene Name
CDC42 effector protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012121.4 1304 Silent Mutation NP_036253.2
XM_005257182.2 1304 Silent Mutation XP_005257239.1

View Full Product Details