Product Details

SNP ID
rs138064774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74367836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGCATGGGGCATCATTGTAGCA[A/G]ACTCAGTCCCCCTCCTCCATCTCTC
Phenotype
MIM: 609046
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTBD17 PubMed Links

Gene Details

Gene
BTBD17
Gene Name
BTB domain containing 17
There are no transcripts associated with this gene.

Gene
GPR142
Gene Name
G protein-coupled receptor 142
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181790.1 Intron NP_861455.1
XM_005257305.3 Intron XP_005257362.1

View Full Product Details