Product Details
- SNP ID
-
rs138385781
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:8803328 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCATCAGAAGGGGCTTGGGCTTG[A/G]CTTTGATTGCTTCCACCTCCTGTTG
- Phenotype
-
MIM: 611462
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MFSD6L
PubMed Links
Gene Details
- Gene
- MFSD6L
- Gene Name
- major facilitator superfamily domain containing 6 like
There are no transcripts associated with this gene.
- Gene
- PIK3R6
- Gene Name
- phosphoinositide-3-kinase regulatory subunit 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001010855.3 |
2032 |
Missense Mutation |
GCC,GTC |
A737V |
NP_001010855.1 |
NM_001290211.1 |
2032 |
Missense Mutation |
GCC,GTC |
A601V |
NP_001277140.1 |
XM_011523677.2 |
2032 |
Silent Mutation |
AGC,AGT |
S697S |
XP_011521979.1 |
XM_011523678.2 |
2032 |
Silent Mutation |
AGC,AGT |
S693S |
XP_011521980.1 |
XM_011523680.2 |
2032 |
Silent Mutation |
AGC,AGT |
S657S |
XP_011521982.1 |
XM_011523681.2 |
2032 |
Silent Mutation |
AGC,AGT |
S618S |
XP_011521983.1 |
XM_011523683.2 |
2032 |
Silent Mutation |
AGC,AGT |
S618S |
XP_011521985.1 |
XM_011523686.2 |
2032 |
Silent Mutation |
AGC,AGT |
S409S |
XP_011521988.1 |
XM_017024228.1 |
2032 |
Silent Mutation |
AGC,AGT |
S698S |
XP_016879717.1 |
XM_017024229.1 |
2032 |
Missense Mutation |
GCC,GTC |
A737V |
XP_016879718.1 |
XM_017024230.1 |
2032 |
Missense Mutation |
GCC,GTC |
A696V |
XP_016879719.1 |
XM_017024231.1 |
2032 |
Silent Mutation |
AGC,AGT |
S618S |
XP_016879720.1 |
View Full Product Details